Product Details

SNP ID
rs8113930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:13785595 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGTTAGGAGGACCCAAATCATC[A/G]GGGCCTTGCAGGATAGGGACTTGGG
Phenotype
MIM: 612360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESF1 PubMed Links

Gene Details

Gene
ESF1
Gene Name
ESF1 nucleolar pre-rRNA processing protein homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276380.1 Intron NP_001263309.1
NM_016649.3 Intron NP_057733.2
XM_017027874.1 Intron XP_016883363.1
Gene
NDUFAF5
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039375.2 Intron NP_001034464.1
NM_024120.4 Intron NP_077025.2
XM_006723620.3 Intron XP_006723683.1
XM_006723622.2 Intron XP_006723685.1
XM_006723623.2 Intron XP_006723686.1
XM_006723624.2 Intron XP_006723687.1
XM_011529342.2 Intron XP_011527644.1
XM_011529344.2 Intron XP_011527646.1
XM_017028061.1 Intron XP_016883550.1

View Full Product Details