Product Details

SNP ID
rs11557574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18142801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTCGAATCCGAGGATCAGGCATC[A/G]GTGGACTTATCGCACGACCAGAGTG
Phenotype
MIM: 614771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAT14 PubMed Links

Gene Details

Gene
KAT14
Gene Name
lysine acetyltransferase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020536.4 578 Silent Mutation TCA,TCG S47S NP_065397.3
Gene
PET117
Gene Name
PET117 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164811.1 578 UTR 3 NP_001158283.1

View Full Product Details