Product Details

SNP ID
rs11544794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33546287 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAACCAGGCGGGTGATGCAGCTG[C/T]CTCTGCCAGGAGTCTACCAAATGAA
Phenotype
MIM: 138440 MIM: 182465
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GART PubMed Links

Gene Details

Gene
GART
Gene Name
phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase
There are no transcripts associated with this gene.

Gene
MIR6501
Gene Name
microRNA 6501
There are no transcripts associated with this gene.

Gene
SON
Gene Name
SON DNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291411.1 207 Missense Mutation GCC,GTC A51V NP_001278340.1
NM_001291412.1 207 Missense Mutation GCC,GTC A51V NP_001278341.1
NM_032195.2 207 Missense Mutation GCC,GTC A51V NP_115571.2
NM_138927.2 207 Missense Mutation GCC,GTC A51V NP_620305.2

View Full Product Details