Product Details

SNP ID
rs4893160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:91918795 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATGGGTTTGTGTGTGCAGTTTT[A/T]AAAAAATTTTTATGAGCACATGATA
Phenotype
MIM: 300246
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDH11X PubMed Links

Gene Details

Gene
PCDH11X
Gene Name
protocadherin 11 X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168360.1 Intron NP_001161832.1
NM_001168361.1 Intron NP_001161833.1
NM_001168362.1 Intron NP_001161834.1
NM_001168363.1 Intron NP_001161835.1
NM_032968.4 Intron NP_116750.1
NM_032969.4 Intron NP_116751.1
XM_011530910.1 Intron XP_011529212.1
XM_011530911.2 Intron XP_011529213.1
XM_011530912.2 Intron XP_011529214.1
XM_011530913.2 Intron XP_011529215.1
XM_011530914.2 Intron XP_011529216.1
XM_011530915.2 Intron XP_011529217.1
XM_011530916.2 Intron XP_011529218.1
XM_017029416.1 Intron XP_016884905.1
XM_017029417.1 Intron XP_016884906.1
XM_017029418.1 Intron XP_016884907.1
XM_017029419.1 Intron XP_016884908.1
XM_017029420.1 Intron XP_016884909.1
XM_017029421.1 Intron XP_016884910.1
XM_017029422.1 Intron XP_016884911.1

View Full Product Details