Product Details

SNP ID
rs7056097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:80676020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAAAACTGTAGTATAAATATAT[A/C]CTTCAGTTCAGTACAAGTCATGAAA
Phenotype
MIM: 300553
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BRWD3 PubMed Links

Gene Details

Gene
BRWD3
Gene Name
bromodomain and WD repeat domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153252.4 6261 UTR 3 NP_694984.4
XM_005262113.3 6261 UTR 3 XP_005262170.1
XM_017029384.1 6261 Intron XP_016884873.1
XM_017029385.1 6261 Intron XP_016884874.1

View Full Product Details