Product Details

SNP ID
rs6654314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136502532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTACATACGACGATATTGGGG[C/T]TCAGGAAGCTTGAATAATTTACCCA
Phenotype
MIM: 300346
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTATSF1 PubMed Links

Gene Details

Gene
HTATSF1
Gene Name
HIV-1 Tat specific factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163280.1 Intron NP_001156752.1
NM_014500.4 Intron NP_055315.2
XM_005262404.3 Intron XP_005262461.1

View Full Product Details