Product Details

SNP ID
rs11099960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154563097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATTGCTACTATTGTGTGTTTTT[C/T]TAGTTAAGTCCCAAGGTGTCAACGA
Phenotype
MIM: 134830
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGB PubMed Links

Gene Details

Gene
FGB
Gene Name
fibrinogen beta chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184741.1 118 Silent Mutation CTA,TTA L27L NP_001171670.1
NM_005141.4 118 Silent Mutation CTA,TTA L27L NP_005132.2

View Full Product Details