Product Details

SNP ID
rs1065410
Assay Type
Functionally Tested
NCBI dbSNP Submissions
7
Location
Chr.1:109688217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGAATACACAGACTCAAGCTA[C/T]GAGGAAAAGAAGTACACGATGGGGG
Phenotype
MIM: 138350 MIM: 138380
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GSTM1 PubMed Links

Gene Details

Gene
GSTM1
Gene Name
glutathione S-transferase mu 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000561.3 Intron NP_000552.2
NM_146421.2 Intron NP_666533.1
XM_005270782.4 Intron XP_005270839.1
Gene
GSTM2
Gene Name
glutathione S-transferase mu 2 (muscle)
There are no transcripts associated with this gene.

View Full Product Details