Product Details

SNP ID
rs4762649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95950937 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGTCTGCTACATATATTTGACGT[C/T]TAAAGTTTTTTTAATTGTAAATTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMDHD1 PubMed Links

Gene Details

Gene
AMDHD1
Gene Name
amidohydrolase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152435.2 Intron NP_689648.2
Gene
CCDC38
Gene Name
coiled-coil domain containing 38
There are no transcripts associated with this gene.

View Full Product Details