Product Details

SNP ID
rs2609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:128094186 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAATTGTAAACATAGCTAATAGC[A/G]TAATACTGCTAGACAGTTCATATGA
Phenotype
MIM: 605897
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UGGT1 PubMed Links

Gene Details

Gene
UGGT1
Gene Name
UDP-glucose glycoprotein glucosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020120.3 Intron NP_064505.1
XM_006712634.3 Intron XP_006712697.1
XM_006712635.3 Intron XP_006712698.1
XM_006712636.3 Intron XP_006712699.1
XM_017004507.1 Intron XP_016859996.1
XM_017004508.1 Intron XP_016859997.1
XM_017004509.1 Intron XP_016859998.1
XM_017004510.1 Intron XP_016859999.1

View Full Product Details