Product Details

SNP ID
rs11556520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55523759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGGTTACCTCAAAGAAATTGG[A/G]ATTAATGAAGATCAATTTCAAGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP36 PubMed Links

Gene Details

Gene
CFAP36
Gene Name
cilia and flagella associated protein 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282761.1 417 Silent Mutation GGA,GGG G98G NP_001269690.1
NM_080667.6 417 Silent Mutation GGA,GGG G73G NP_542398.3
XM_011532499.1 417 Intron XP_011530801.1
XM_017003254.1 417 UTR 5 XP_016858743.1
XM_017003255.1 417 UTR 5 XP_016858744.1

View Full Product Details