Product Details

SNP ID
rs11545479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70088167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGGGGATATGCTGCCCAACTCC[A/T]CCGAGCGGGCCATCACCATCGCTGG
Phenotype
MIM: 601209
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PCBP1 PubMed Links

Gene Details

Gene
PCBP1
Gene Name
poly(rC) binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006196.3 715 Missense Mutation ACC,TCC T142S NP_006187.2
Gene
PCBP1-AS1
Gene Name
PCBP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details