Product Details

SNP ID
rs7557705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73928514 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTGAAGATGAGGGAGCAAATCAC[G/T]TGGATACCTGGGAGAAGAATCTTCT
Phenotype
MIM: 102545 MIM: 601465
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACTG2 PubMed Links

Gene Details

Gene
ACTG2
Gene Name
actin, gamma 2, smooth muscle, enteric
There are no transcripts associated with this gene.

Gene
DGUOK
Gene Name
deoxyguanosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318859.1 Intron NP_001305788.1
NM_001318860.1 Intron NP_001305789.1
NM_001318861.1 Intron NP_001305790.1
NM_001318862.1 Intron NP_001305791.1
NM_001318863.1 Intron NP_001305792.1
NM_080916.2 Intron NP_550438.1
NM_080918.2 Intron NP_550440.1
XM_011532647.2 Intron XP_011530949.1

View Full Product Details