Product Details

SNP ID
hCV44349210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85362985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCAACCCATCAGGGCACACTA[A/G]GTCCTTTTGGCCTCCAGTGTCAAGG
Phenotype
MIM: 615427
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELMOD3 PubMed Links

Gene Details

Gene
ELMOD3
Gene Name
ELMO domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135021.1 Intron NP_001128493.1
NM_001135022.1 Intron NP_001128494.1
NM_001135023.1 Intron NP_001128495.1
Gene
RETSAT
Gene Name
retinol saturase
There are no transcripts associated with this gene.

View Full Product Details