Product Details

SNP ID
rs7610463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:72866257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCGTGACAAGAACCAGTGACTGA[C/T]GAACAACTGAGTTCACCAGTTCACT
Phenotype
MIM: 613322
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GXYLT2 PubMed Links

Gene Details

Gene
GXYLT2
Gene Name
glucoside xylosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080393.1 Intron NP_001073862.1
XM_011534067.2 Intron XP_011532369.1
XM_011534068.1 Intron XP_011532370.1
XM_017007123.1 Intron XP_016862612.1
XM_017007124.1 Intron XP_016862613.1

View Full Product Details