Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001253387.1 | Intron | NP_001240316.1 | |||
NM_001253388.1 | Intron | NP_001240317.1 | |||
NM_006614.3 | Intron | NP_006605.2 | |||
XM_006712938.1 | Intron | XP_006713001.1 | |||
XM_006712939.3 | Intron | XP_006713002.1 | |||
XM_006712940.3 | Intron | XP_006713003.1 | |||
XM_011533292.1 | Intron | XP_011531594.1 | |||
XM_011533294.1 | Intron | XP_011531596.1 | |||
XM_011533295.1 | Intron | XP_011531597.1 | |||
XM_011533296.1 | Intron | XP_011531598.1 | |||
XM_017005566.1 | Intron | XP_016861055.1 | |||
XM_017005567.1 | Intron | XP_016861056.1 | |||
XM_017005568.1 | Intron | XP_016861057.1 | |||
XM_017005569.1 | Intron | XP_016861058.1 | |||
XM_017005570.1 | Intron | XP_016861059.1 | |||
XM_017005571.1 | Intron | XP_016861060.1 | |||
XM_017005572.1 | Intron | XP_016861061.1 | |||
XM_017005573.1 | Intron | XP_016861062.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_017007606.1 | Intron | XP_016863095.1 |