Product Details

SNP ID
rs4676633
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39331436 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTGTTAAGGACACTAGTTCTAT[C/T]AGATTAGGGCCCTGCCTTTTTTGGT
Phenotype
MIM: 601834
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCR8 PubMed Links

Gene Details

Gene
CCR8
Gene Name
C-C motif chemokine receptor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005201.3 Intron NP_005192.1

View Full Product Details