Product Details
- SNP ID
-
rs7621701
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:39410221 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGAATCTTTTTTGCCACAGCTCTA[A/T]TTTAATAATTTTTCTGTTGAGAAAA
- Phenotype
-
MIM: 150370
MIM: 180646
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RPSA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75586456] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RPSA
- Gene Name
- ribosomal protein SA
- Gene
- SNORA6
- Gene Name
- small nucleolar RNA, H/ACA box 6
There are no transcripts associated with this gene.
- Gene
- SNORA62
- Gene Name
- small nucleolar RNA, H/ACA box 62
There are no transcripts associated with this gene.
View Full Product Details