Product Details

SNP ID
rs11559016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42258132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTCATACTCCTCGGCACTGCGA[C/T]GGCCAAAATCCATCCAGCCCATGTA
Phenotype
MIM: 118440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCK PubMed Links

Gene Details

Gene
CCK
Gene Name
cholecystokinin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000729.5 1208 Missense Mutation CAT,CGT H105R NP_000720.1
NM_001174138.2 1208 Missense Mutation CAT,CGT H105R NP_001167609.1

View Full Product Details