Product Details

SNP ID
rs7430833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48868813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTTTATGGAGTTGTAATGAAGT[A/T]CCCAACATGTTACCAGCGTGGAGTC
Phenotype
MIM: 613698
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A20 PubMed Links

Gene Details

Gene
SLC25A20
Gene Name
solute carrier family 25 member 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000387.5 Intron NP_000378.1

View Full Product Details