Product Details

SNP ID
rs7623023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48876805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGAGATTCTGGACAAATGTTA[A/G]AAACACATGGCTGAGCACAGTGGAT
Phenotype
MIM: 613698
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A20 PubMed Links

Gene Details

Gene
SLC25A20
Gene Name
solute carrier family 25 member 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000387.5 Intron NP_000378.1

View Full Product Details