Product Details

SNP ID
rs11538558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51389088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCAGTGCACGGACCGATTTGT[A/G]GTCTGCTCAATCTCTGTTGCACCTG
Phenotype
MIM: 603123 MIM: 601916 MIM: 612602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF1 PubMed Links

Gene Details

Gene
DCAF1
Gene Name
DDB1 and CUL4 associated factor 1
There are no transcripts associated with this gene.

Gene
DOCK3
Gene Name
dedicator of cytokinesis 3
There are no transcripts associated with this gene.

Gene
MANF
Gene Name
mesencephalic astrocyte derived neurotrophic factor
There are no transcripts associated with this gene.

Gene
RBM15B
Gene Name
RNA binding motif protein 15B
There are no transcripts associated with this gene.

View Full Product Details