Product Details

SNP ID
rs5745106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173145270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCCGTTTGAACAGATAGGAAAT[G/T]AAGTGTTTCGCCTTAAGCCATTTAA
Phenotype
MIM: 603291
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BNIP1 PubMed Links

Gene Details

Gene
BNIP1
Gene Name
BCL2 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205.2 Intron NP_001196.2
NM_013978.2 Intron NP_053581.2
NM_013979.2 Intron NP_053582.2
NM_013980.2 Intron NP_053583.2
XM_011534638.1 Intron XP_011532940.1
XM_011534639.1 Intron XP_011532941.1
Gene
CREBRF
Gene Name
CREB3 regulatory factor
There are no transcripts associated with this gene.

View Full Product Details