Product Details

SNP ID
rs2075667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176620760 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGACAGCCCTGGCTCTGGGGGGCG[A/G]GGCAGGGACAGGGACAGAATTGTGC
Phenotype
MIM: 602569
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4E1B PubMed Links

Gene Details

Gene
EIF4E1B
Gene Name
eukaryotic translation initiation factor 4E family member 1B
There are no transcripts associated with this gene.

Gene
MIR4281
Gene Name
microRNA 4281
There are no transcripts associated with this gene.

Gene
SNCB
Gene Name
synuclein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001502.2 747 UTR 3 NP_001001502.1
NM_001318034.1 747 UTR 3 NP_001304963.1
NM_001318035.1 747 UTR 3 NP_001304964.1
NM_001318036.1 747 UTR 3 NP_001304965.1
NM_001318037.1 747 UTR 3 NP_001304966.1
NM_003085.4 747 UTR 3 NP_003076.1
XM_006714914.3 747 Intron XP_006714977.1
XM_006714915.3 747 Intron XP_006714978.1
XM_006714916.2 747 UTR 3 XP_006714979.1
XM_011534640.2 747 Intron XP_011532942.1

View Full Product Details