Product Details

SNP ID
rs923321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176621957 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAGCCACCTGCAGGTGACCACAT[A/G]TATGTGAACAAACATGGGACTGAGC
Phenotype
MIM: 602569
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4E1B PubMed Links

Gene Details

Gene
EIF4E1B
Gene Name
eukaryotic translation initiation factor 4E family member 1B
There are no transcripts associated with this gene.

Gene
MIR4281
Gene Name
microRNA 4281
There are no transcripts associated with this gene.

Gene
SNCB
Gene Name
synuclein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001502.2 Intron NP_001001502.1
NM_001318034.1 Intron NP_001304963.1
NM_001318035.1 Intron NP_001304964.1
NM_001318036.1 Intron NP_001304965.1
NM_001318037.1 Intron NP_001304966.1
NM_003085.4 Intron NP_003076.1
XM_006714914.3 Intron XP_006714977.1
XM_006714915.3 Intron XP_006714978.1
XM_006714916.2 Intron XP_006714979.1
XM_011534640.2 Intron XP_011532942.1

View Full Product Details