Product Details
- SNP ID
-
rs7733295
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:95620641 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCGGCCACGGGGGACCTGGCGTG[C/T]CGCTTCCTGCAGCTGCTGCAGGCAT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GPR150
PubMed Links
Gene Details
- Gene
- GPR150
- Gene Name
- G protein-coupled receptor 150
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_199243.1 |
366 |
Silent Mutation |
TGC,TGT |
C122C |
NP_954713.1 |
View Full Product Details