Product Details

SNP ID
rs11547126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143021708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCAATTTCAGTGCGGCCGATGG[C/T]GGGCTGCGGGCAAGCGTCACCCTGC
Phenotype
MIM: 601384
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100133669 PubMed Links

Gene Details

Gene
LOC100133669
Gene Name
uncharacterized LOC100133669
There are no transcripts associated with this gene.

Gene
LY6E
Gene Name
lymphocyte antigen 6 complex, locus E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127213.1 474 Silent Mutation GGC,GGT G105G NP_001120685.1
NM_002346.2 474 Silent Mutation GGC,GGT G105G NP_002337.1

View Full Product Details