Product Details
- SNP ID
-
rs17739208
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:617000 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAGAGAGACATTGGTTATATAAGC[A/G]AGCAAATATCCAGAAGGAAGAAAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ERICH1
PubMed Links
Gene Details
- Gene
- ERICH1
- Gene Name
- glutamate rich 1
View Full Product Details