Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000718.3 | Intron | NP_000709.1 | |||
NM_001243812.1 | Intron | NP_001230741.1 | |||
XM_011518991.2 | Intron | XP_011517293.1 | |||
XM_011518992.2 | Intron | XP_011517294.1 | |||
XM_011518993.2 | Intron | XP_011517295.1 | |||
XM_011518994.2 | Intron | XP_011517296.1 | |||
XM_017015115.1 | Intron | XP_016870604.1 | |||
XM_017015116.1 | Intron | XP_016870605.1 | |||
XM_017015117.1 | Intron | XP_016870606.1 | |||
XM_017015118.1 | Intron | XP_016870607.1 | |||
XM_017015119.1 | Intron | XP_016870608.1 | |||
XM_017015120.1 | Intron | XP_016870609.1 |