Product Details

SNP ID
rs7331172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:25161449 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGAGTAAAAGACTCAGAATTCA[A/G]ATCAGAGTCGGAATAACAAATGAAC
Phenotype
MIM: 614659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMER2 PubMed Links

Gene Details

Gene
AMER2
Gene Name
APC membrane recruitment protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152704.3 Intron NP_689917.2
NM_199138.2 Intron NP_954589.1
XM_005266279.3 Intron XP_005266336.1
XM_005266280.3 Intron XP_005266337.1
XM_017020415.1 Intron XP_016875904.1

View Full Product Details