Product Details
- SNP ID
-
hCV44655727
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:56606665 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGGGGAAAAGGAAAAGTCACCTA[A/G]CTGGTAACGTCTGAGAATACTCTGT
- Phenotype
-
MIM: 603049
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TXNL1
PubMed Links
Gene Details
- Gene
- TXNL1
- Gene Name
- thioredoxin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004786.2 |
|
Intron |
|
|
NP_004777.1 |
View Full Product Details