Product Details

SNP ID
rs11547601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:142147321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGCACCGCTGCCCCCGCTCCCC[A/G]CACAGTTCAAGAGCATACAGCATCA
Phenotype
MIM: 162341 MIM: 610902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NMBR PubMed Links

Gene Details

Gene
NMBR
Gene Name
neuromedin B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324307.1 160 Intron NP_001311236.1
NM_001324308.1 160 Intron NP_001311237.1
NM_002511.3 160 Intron NP_002502.2
XM_011535846.1 160 Intron XP_011534148.1
XM_017010901.1 160 Intron XP_016866390.1
XM_017010902.1 160 Intron XP_016866391.1
Gene
VTA1
Gene Name
vesicle trafficking 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286371.1 160 Missense Mutation ACA,GCA T12A NP_001273300.1
NM_001286372.1 160 UTR 5 NP_001273301.1
NM_016485.4 160 Missense Mutation ACA,GCA T12A NP_057569.2

View Full Product Details