Product Details

SNP ID
rs8021588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:33924643 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGGATTTTTAAAACATGAAAGT[A/G]ATACTGAAAACTAGGAAAAGTAGCT
Phenotype
MIM: 606426
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGLN3 PubMed Links

Gene Details

Gene
EGLN3
Gene Name
egl-9 family hypoxia inducible factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308103.1 1760 UTR 3 NP_001295032.1
NM_022073.3 1760 UTR 3 NP_071356.1

View Full Product Details