Product Details

SNP ID
rs11538184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24440870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTACGAGGTTGGAGAAAAAGTCAA[A/T]GTCCCCTCCTGGGCTGTCTTCATCA
Phenotype
MIM: 616162
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KHNYN PubMed Links

Gene Details

Gene
KHNYN
Gene Name
KH and NYN domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290256.1 377 UTR 3 NP_001277185.1
NM_001290257.1 377 UTR 3 NP_001277186.1
NM_015299.2 377 UTR 3 NP_056114.1
XM_005267474.4 377 UTR 3 XP_005267531.1
XM_011536590.2 377 UTR 3 XP_011534892.1
XM_017021114.1 377 Intron XP_016876603.1
Gene
LOC101927045
Gene Name
uncharacterized LOC101927045
There are no transcripts associated with this gene.

Gene
SDR39U1
Gene Name
short chain dehydrogenase/reductase family 39U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290292.1 377 Missense Mutation ATT,TTT I47F NP_001277221.1
NM_001290293.2 377 Missense Mutation ATT,TTT I21F NP_001277222.1
NM_001308075.1 377 Missense Mutation ATT,TTT I47F NP_001295004.1
NM_020195.2 377 Missense Mutation ATT,TTT I129F NP_064580.2
XM_011536975.1 377 Missense Mutation ATT,TTT I152F XP_011535277.1
XM_011536976.1 377 Missense Mutation ATT,TTT I87F XP_011535278.1

View Full Product Details