Product Details

SNP ID
rs8019781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96808264 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAAATGAATAGGACATGGTCCC[C/T]GCTTTCTGCCCCTGTTTTTGTTTAA
Phenotype
MIM: 602168
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VRK1 PubMed Links

Gene Details

Gene
VRK1
Gene Name
vaccinia related kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003384.2 Intron NP_003375.1
XM_006720247.3 Intron XP_006720310.1
XM_011537132.1 Intron XP_011535434.1
XM_017021622.1 Intron XP_016877111.1
XM_017021624.1 Intron XP_016877113.1
XM_017021625.1 Intron XP_016877114.1
XM_017021626.1 Intron XP_016877115.1

View Full Product Details