Product Details

SNP ID
rs55633823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105488368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTACTGCAACCACCCCTGCTACG[C/T]AGCCATGTTTGGGCCTAAAGGTATG
Phenotype
MIM: 123875 MIM: 601183
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf80 PubMed Links

Gene Details

Gene
C14orf80
Gene Name
chromosome 14 open reading frame 80
There are no transcripts associated with this gene.

Gene
CRIP1
Gene Name
cysteine rich protein 1
There are no transcripts associated with this gene.

Gene
CRIP2
Gene Name
cysteine rich protein 2
There are no transcripts associated with this gene.

View Full Product Details