Product Details

SNP ID
rs10147883
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105632009 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACTCTTAGTCCACAATCTCAGAG[A/G]AATATCACCTGCATCAAGTGTGGGA
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010 MIM: 147130
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

Gene
IGHG4
Gene Name
immunoglobulin heavy constant gamma 4 (G4m marker)
There are no transcripts associated with this gene.

Gene
MIR8071-2
Gene Name
microRNA 8071-2
There are no transcripts associated with this gene.

View Full Product Details