Product Details

SNP ID
rs4778415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:24168193 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGATCAGTCATTTTCTGGGGAT[C/T]CATTGTATAAAGTGGGGTGGAGTTT
Phenotype
MIM: 611217
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105370733 PubMed Links

Gene Details

Gene
LOC105370733
Gene Name
uncharacterized LOC105370733
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017022805.1 Intron XP_016878294.1
XM_017022806.1 Intron XP_016878295.1
Gene
PWRN2
Gene Name
Prader-Willi region non-protein coding RNA 2
There are no transcripts associated with this gene.

View Full Product Details