Product Details

SNP ID
rs12995235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96817191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCGACCCCTACAGCGATCTCA[G/T]CAAGGGCGTGCTGCGCTGCCGGACC
Phenotype
MIM: 607804 MIM: 607805
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNNM3 PubMed Links

Gene Details

Gene
CNNM3
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017623.4 938 Missense Mutation AGC,ATC S305I NP_060093.3
NM_199078.2 938 Missense Mutation AGC,ATC S305I NP_951060.1
XM_011510957.2 938 Missense Mutation AGC,ATC S305I XP_011509259.1
XM_017003800.1 938 Missense Mutation AGC,ATC S305I XP_016859289.1
Gene
CNNM4
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
There are no transcripts associated with this gene.

View Full Product Details