Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_138492.5 | Intron | NP_612501.3 | |||
NM_182960.3 | Intron | NP_892005.1 | |||
NM_205846.2 | Intron | NP_995318.1 | |||
XM_017009127.1 | Intron | XP_016864616.1 | |||
XM_017009128.1 | Intron | XP_016864617.1 | |||
XM_017009129.1 | Intron | XP_016864618.1 | |||
XM_017009130.1 | Intron | XP_016864619.1 | |||
XM_017009131.1 | Intron | XP_016864620.1 | |||
XM_017009132.1 | Intron | XP_016864621.1 | |||
XM_017009133.1 | Intron | XP_016864622.1 | |||
XM_017009134.1 | Intron | XP_016864623.1 | |||
XM_017009135.1 | Intron | XP_016864624.1 |