Product Details

SNP ID
rs7980901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50523956 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGCCTCTTCCTGCAATGTCTTT[C/T]CAGTGCCCTCTATGGACAGGCAAAG
Phenotype
MIM: 611379
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIP2B PubMed Links

Gene Details

Gene
DIP2B
Gene Name
disco interacting protein 2 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173602.2 Intron NP_775873.2
XM_005269044.1 Intron XP_005269101.1
XM_006719520.3 Intron XP_006719583.1
XM_006719521.3 Intron XP_006719584.1
XM_011538594.2 Intron XP_011536896.1
XM_011538595.2 Intron XP_011536897.1
XM_011538596.2 Intron XP_011536898.1
XM_017019699.1 Intron XP_016875188.1
XM_017019700.1 Intron XP_016875189.1
XM_017019701.1 Intron XP_016875190.1
XM_017019702.1 Intron XP_016875191.1
XM_017019703.1 Intron XP_016875192.1
XM_017019704.1 Intron XP_016875193.1
XM_017019705.1 Intron XP_016875194.1

View Full Product Details