Product Details
- SNP ID
-
rs7301225
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:54644920 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGATGGAGGTTAGATTCACAGGA[G/T]CCCCAAAGACCAACCTCTCTTCCCC
- Phenotype
-
MIM: 606634
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DCD
PubMed Links
Gene Details
- Gene
- DCD
- Gene Name
- dermcidin
View Full Product Details