Product Details

SNP ID
rs7975515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68618683 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTCTGGCGCTTTAGAATAAACCA[C/G]GTTAGAGTACTTCACTGTCCTAGTA
Phenotype
MIM: 179530
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAP1B PubMed Links

Gene Details

Gene
RAP1B
Gene Name
RAP1B, member of RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010942.2 Intron NP_001010942.1
NM_001251917.1 Intron NP_001238846.1
NM_001251918.1 Intron NP_001238847.1
NM_001251921.1 Intron NP_001238850.1
NM_001251922.1 Intron NP_001238851.1
NM_015646.5 Intron NP_056461.1
Gene
SNORA70G
Gene Name
small nucleolar RNA, H/ACA box 70G
There are no transcripts associated with this gene.

View Full Product Details