Product Details
- SNP ID
-
rs4913758
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:10414849 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCTGCCCAGCTGCTCCAAGCCAG[G/A]CTGATGAAGGAGGAGTCCCCTGTGG
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BAGE2
PubMed Links
Gene Details
- Gene
- BAGE2
- Gene Name
- B melanoma antigen family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182482.2 |
262 |
Silent Mutation |
AGA,AGG |
R18R |
NP_872288.2 |
View Full Product Details