Product Details
- SNP ID
-
rs12631343
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:197051652 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTTTCTGGCTTGTTCTTCTGTTA[C/G]ACGCTTATTCATTTCCCTACGAAAA
- Phenotype
-
MIM: 601014
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DLG1
PubMed Links
Gene Details
- Gene
- DLG1
- Gene Name
- discs large MAGUK scaffold protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098424.1 |
2676 |
Missense Mutation |
CTA,GTA |
L845V |
NP_001091894.1 |
NM_001204386.1 |
2676 |
Missense Mutation |
CTA,GTA |
L833V |
NP_001191315.1 |
NM_001204387.1 |
2676 |
Missense Mutation |
CTA,GTA |
L741V |
NP_001191316.1 |
NM_001204388.1 |
2676 |
Missense Mutation |
CTA,GTA |
L729V |
NP_001191317.1 |
NM_001290983.1 |
2676 |
Missense Mutation |
CTA,GTA |
L845V |
NP_001277912.1 |
NM_004087.2 |
2676 |
Missense Mutation |
CTA,GTA |
L867V |
NP_004078.2 |
XM_005269289.3 |
2676 |
Missense Mutation |
CTA,GTA |
L867V |
XP_005269346.1 |
XM_011512502.2 |
2676 |
Missense Mutation |
CTA,GTA |
L845V |
XP_011510804.1 |
XM_011512503.1 |
2676 |
Missense Mutation |
CTA,GTA |
L833V |
XP_011510805.1 |
XM_011512505.1 |
2676 |
Missense Mutation |
CTA,GTA |
L812V |
XP_011510807.1 |
XM_011512506.1 |
2676 |
Missense Mutation |
CTA,GTA |
L794V |
XP_011510808.1 |
XM_011512509.1 |
2676 |
Intron |
|
|
XP_011510811.1 |
XM_017005800.1 |
2676 |
Missense Mutation |
CTA,GTA |
L867V |
XP_016861289.1 |
XM_017005801.1 |
2676 |
Missense Mutation |
CTA,GTA |
L867V |
XP_016861290.1 |
XM_017005802.1 |
2676 |
Missense Mutation |
CTA,GTA |
L867V |
XP_016861291.1 |
XM_017005803.1 |
2676 |
Missense Mutation |
CTA,GTA |
L867V |
XP_016861292.1 |
XM_017005804.1 |
2676 |
Missense Mutation |
CTA,GTA |
L866V |
XP_016861293.1 |
XM_017005805.1 |
2676 |
Missense Mutation |
CTA,GTA |
L845V |
XP_016861294.1 |
XM_017005806.1 |
2676 |
Missense Mutation |
CTA,GTA |
L834V |
XP_016861295.1 |
XM_017005807.1 |
2676 |
Missense Mutation |
CTA,GTA |
L834V |
XP_016861296.1 |
XM_017005808.1 |
2676 |
Missense Mutation |
CTA,GTA |
L834V |
XP_016861297.1 |
XM_017005809.1 |
2676 |
Missense Mutation |
CTA,GTA |
L834V |
XP_016861298.1 |
XM_017005810.1 |
2676 |
Missense Mutation |
CTA,GTA |
L833V |
XP_016861299.1 |
XM_017005811.1 |
2676 |
Missense Mutation |
CTA,GTA |
L816V |
XP_016861300.1 |
XM_017005812.1 |
2676 |
Missense Mutation |
CTA,GTA |
L816V |
XP_016861301.1 |
XM_017005813.1 |
2676 |
Missense Mutation |
CTA,GTA |
L816V |
XP_016861302.1 |
XM_017005814.1 |
2676 |
Missense Mutation |
CTA,GTA |
L816V |
XP_016861303.1 |
XM_017005815.1 |
2676 |
Missense Mutation |
CTA,GTA |
L815V |
XP_016861304.1 |
XM_017005816.1 |
2676 |
Missense Mutation |
CTA,GTA |
L812V |
XP_016861305.1 |
XM_017005817.1 |
2676 |
Missense Mutation |
CTA,GTA |
L812V |
XP_016861306.1 |
XM_017005818.1 |
2676 |
Missense Mutation |
CTA,GTA |
L812V |
XP_016861307.1 |
XM_017005819.1 |
2676 |
Missense Mutation |
CTA,GTA |
L812V |
XP_016861308.1 |
XM_017005820.1 |
2676 |
Missense Mutation |
CTA,GTA |
L794V |
XP_016861309.1 |
XM_017005821.1 |
2676 |
Missense Mutation |
CTA,GTA |
L784V |
XP_016861310.1 |
XM_017005822.1 |
2676 |
Missense Mutation |
CTA,GTA |
L762V |
XP_016861311.1 |
XM_017005823.1 |
2676 |
Missense Mutation |
CTA,GTA |
L751V |
XP_016861312.1 |
XM_017005824.1 |
2676 |
Missense Mutation |
CTA,GTA |
L460V |
XP_016861313.1 |
View Full Product Details