Product Details

SNP ID
rs7098317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97742588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGGTATTCAAAATAAACTCTCA[A/G]ATAAACTCTCAAATTTTCAAAAACA
Phenotype
MIM: 610243
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFYVE27 PubMed Links

Gene Details

Gene
ZFYVE27
Gene Name
zinc finger FYVE-type containing 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002261.3 Intron NP_001002261.1
NM_001002262.3 Intron NP_001002262.1
NM_001174119.1 Intron NP_001167590.1
NM_001174120.1 Intron NP_001167591.1
NM_001174121.1 Intron NP_001167592.1
NM_001174122.1 Intron NP_001167593.1
NM_144588.6 Intron NP_653189.3
XM_005269502.3 Intron XP_005269559.1
XM_005269503.3 Intron XP_005269560.1
XM_005269504.3 Intron XP_005269561.1
XM_005269505.4 Intron XP_005269562.1
XM_005269506.3 Intron XP_005269563.1
XM_005269508.3 Intron XP_005269565.1
XM_005269509.3 Intron XP_005269566.1
XM_005269510.3 Intron XP_005269567.1
XM_005269511.3 Intron XP_005269568.1
XM_011539252.2 Intron XP_011537554.1
XM_011539253.2 Intron XP_011537555.1
XM_011539254.2 Intron XP_011537556.1
XM_011539255.2 Intron XP_011537557.1
XM_017015644.1 Intron XP_016871133.1
XM_017015645.1 Intron XP_016871134.1
XM_017015646.1 Intron XP_016871135.1
XM_017015647.1 Intron XP_016871136.1
XM_017015648.1 Intron XP_016871137.1
XM_017015649.1 Intron XP_016871138.1
XM_017015650.1 Intron XP_016871139.1
XM_017015651.1 Intron XP_016871140.1
XM_017015652.1 Intron XP_016871141.1
XM_017015653.1 Intron XP_016871142.1
XM_017015654.1 Intron XP_016871143.1
XM_017015655.1 Intron XP_016871144.1

View Full Product Details