Product Details

SNP ID
hCV44902356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103392226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTCACAGCTGGAGTTTTTTTGG[A/G]CCTTAACTTGAAATATAAGACAATC
Phenotype
MIM: 612333 MIM: 601787 MIM: 615204
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1307 PubMed Links

Gene Details

Gene
MIR1307
Gene Name
microRNA 1307
There are no transcripts associated with this gene.

Gene
PDCD11
Gene Name
programmed cell death 11
There are no transcripts associated with this gene.

Gene
TAF5
Gene Name
TATA-box binding protein associated factor 5
There are no transcripts associated with this gene.

Gene
USMG5
Gene Name
up-regulated during skeletal muscle growth 5 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206426.1 455 Intron NP_001193355.1
NM_001206427.1 455 Missense Mutation CCC,TCC P49S NP_001193356.1
NM_032747.3 455 Missense Mutation CCC,TCC P49S NP_116136.1

View Full Product Details