Product Details

SNP ID
rs6997277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:20146470 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCACCACTCCCTATCACACCTTC[A/T]TGTCTTCTTTTGTTCCCTTGAGCAG
Phenotype
MIM: 193002
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC18A1 PubMed Links

Gene Details

Gene
SLC18A1
Gene Name
solute carrier family 18 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135691.2 Intron NP_001129163.1
NM_001142324.1 Intron NP_001135796.1
NM_001142325.1 Intron NP_001135797.1
NM_003053.3 Intron NP_003044.1
XM_011544625.1 Intron XP_011542927.1
XM_011544626.1 Intron XP_011542928.1
XM_011544627.1 Intron XP_011542929.1

View Full Product Details