Product Details

SNP ID
rs12564635
Assay Type
Functionally tested
NCBI dbSNP Submissions
58
Location
Chr.1:47094367 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAACATCCTTATTTGAAGGAAGA[G/T]AAATTGTGGCTAAAAGGTGGGATGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP4Z1 PubMed Links

Gene Details

Gene
CYP4Z1
Gene Name
cytochrome P450 family 4 subfamily Z member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178134.2 Intron NP_835235.1

View Full Product Details