Product Details

SNP ID
rs5744279
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112145874 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAGAGGTAAGGACATCCTTGGGC[A/G]GTAGCTTTCCTAGTTTTGATGTTGA
Phenotype
MIM: 600953
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IL18 PubMed Links

Gene Details

Gene
IL18
Gene Name
interleukin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243211.1 Intron NP_001230140.1
NM_001562.3 Intron NP_001553.1
XM_011542805.1 Intron XP_011541107.1
XM_011542806.2 Intron XP_011541108.1

View Full Product Details